ENA341291460 Enamine

2-{[(4-tert-butylphenyl)methyl]sulfanyl}acetic acid

  • MDL number MFCD08444798
  • Cas Number : 17874-35-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86691

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86691
Availability details for
Quantity
  • Availability
  • Add to Cart