ENA341291463 Enamine

3-methyl-1-[4-(trifluoromethoxy)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD03034587
  • Cas Number : 497141-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86694

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86694
Availability details for
Quantity
  • Availability
  • Add to Cart