ENA341291479 Enamine

(2R)-2-({[(4-methylphenyl)amino]carbonyl}amino)propanoic acid

  • MDL number MFCD12169864
  • Cas Number : 1308987-90-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86710

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86710
Availability details for
Quantity
  • Availability
  • Add to Cart