ENA341291567 Enamine

1-isonicotinoylpiperidine-3-carboxylic acid

  • MDL number MFCD08444920
  • Cas Number : 67691-63-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 258...260
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86799

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86799
Availability details for
Quantity
  • Availability
  • Add to Cart