ENA341291580 Enamine

1-(3,4-dimethoxyphenyl)-2,5-dimethyl-1H-pyrrole-3-carbaldehyde

  • MDL number MFCD08444931
  • Cas Number : 923772-28-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86812
Availability details for
Quantity
  • Availability
  • Add to Cart