ENA341291611 Enamine

5,7-dimethyl-2-phenylquinoline-4-carboxylic acid

  • MDL number MFCD04035073
  • Cas Number : 436089-00-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86844

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86844
Availability details for
Quantity
  • Availability
  • Add to Cart