ENA341291631 Enamine

1-isonicotinoylpyrrolidine-2-carboxylic acid

  • MDL number MFCD00448260

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 79...81
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86864

Safety Information

msds https://www.enaminestore.com/catalog/EN300-86864
Availability details for
Quantity
  • Availability
  • Add to Cart