ENA341291645 Enamine

2-(prop-2-yn-1-ylsulfanyl)acetic acid

  • MDL number MFCD08445003
  • Cas Number : 13702-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-86878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86878
Availability details for
Quantity
  • Availability
  • Add to Cart