ENA341291666 Enamine

2-cyclopropyl-7-fluoroquinoline-4-carboxylic acid

  • MDL number MFCD08445029
  • Cas Number : 923751-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-86899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86899
Availability details for
Quantity
  • Availability
  • Add to Cart