ENA341291725 Enamine

3-(piperidine-1-sulfonyl)benzene-1-sulfonyl chloride

  • MDL number MFCD08445100
  • Cas Number : 920477-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-86961

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-86961
Availability details for
Quantity
  • Availability
  • Add to Cart