ENA341291808 Enamine

1-(4-methoxybenzoyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD00441408
  • Cas Number : 62522-92-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87045

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87045
Availability details for
Quantity
  • Availability
  • Add to Cart