ENA341291855 Enamine

methyl 2-methyl-4H-pyrrolo[3,2-d][1,3]thiazole-5-carboxylate

  • MDL number MFCD09297331
  • Cas Number : 238749-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87092

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87092
Availability details for
Quantity
  • Availability
  • Add to Cart