ENA341291879 Enamine

1-(1-methyl-1H-pyrazol-4-yl)-2-(piperazin-1-yl)ethan-1-one dihydrochloride

  • MDL number MFCD20441581
  • Cas Number : 1354951-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-87142

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87142
Availability details for
Quantity
  • Availability
  • Add to Cart