ENA341291884 Enamine

6-fluoro-8-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD12192835
  • Cas Number : 1065094-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-87147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87147
Availability details for
Quantity
  • Availability
  • Add to Cart