ENA341291916 Enamine

methyl 6-[(4-methylbenzenesulfonyl)oxy]-4-(trifluoromethyl)thieno[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD03656278
  • Cas Number : 633315-51-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87182

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87182
Availability details for
Quantity
  • Availability
  • Add to Cart