ENA341291938 Enamine

2-(5-oxopyrrolidin-2-yl)acetonitrile

  • MDL number MFCD09040736
  • Cas Number : 98134-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87205

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87205
Availability details for
Quantity
  • Availability
  • Add to Cart