ENA341291983 Enamine

6-methoxy-4-(trifluoromethyl)quinoline-3-carboxylic acid

  • MDL number MFCD09040801
  • Cas Number : 929974-06-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87263

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87263
Availability details for
Quantity
  • Availability
  • Add to Cart