ENA341291994 Enamine

2-(3-methyl-1,2,4-oxadiazol-5-yl)benzoic acid

  • MDL number MFCD08741426
  • Cas Number : 475105-77-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-87274

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87274
Availability details for
Quantity
  • Availability
  • Add to Cart