ENA341291996 Enamine

4-(3-bromopropyl)morpholine hydrobromide

  • MDL number MFCD13176498
  • Cas Number : 88806-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87276

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87276
Availability details for
Quantity
  • Availability
  • Add to Cart