ENA341292016 Enamine

3-amino-1-oxo-1H,5H,10H,10aH-pyrrolo[1,2-b]isoquinoline-2-carbonitrile

  • MDL number MFCD09702221
  • Cas Number : 929974-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87297

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87297
Availability details for
Quantity
  • Availability
  • Add to Cart