ENA341292083 Enamine

3-[(1-methyl-1H-pyrazol-4-yl)amino]benzoic acid

  • MDL number MFCD20501923
  • Cas Number : 1354962-77-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87369
Availability details for
Quantity
  • Availability
  • Add to Cart