ENA341292086 Enamine

4-(4-fluorophenyl)-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD09746413
  • Cas Number : 191668-31-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 199...201
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87372

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87372
Availability details for
Quantity
  • Availability
  • Add to Cart