ENA341292089 Enamine

6-(2,3-dichlorophenoxy)pyridazine-3-carboxylic acid

  • MDL number MFCD12722739
  • Cas Number : 1184088-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87375

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87375
Availability details for
Quantity
  • Availability
  • Add to Cart