ENA341292090 Enamine

5-[6-fluoro-1-(propan-2-yl)-1H-1,3-benzodiazol-2-yl]-2-methylaniline

  • MDL number MFCD20441585
  • Cas Number : 1333549-25-9
  • Molecular Weight : 283.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-87376

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87376
Availability details for
Quantity
  • Availability
  • Add to Cart