ENA341292121 Enamine

2-(chloromethyl)-5,6-dimethylfuro[2,3-d]pyrimidin-4-amine

  • MDL number MFCD09702320
  • Cas Number : 1000933-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87423

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87423
Availability details for
Quantity
  • Availability
  • Add to Cart