ENA341292129 Enamine

6-chloropyridazine-3-sulfonyl fluoride

  • MDL number MFCD09702326
  • Cas Number : 209848-48-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87431

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87431
Availability details for
Quantity
  • Availability
  • Add to Cart