ENA341292142 Enamine

1-(3,5-dimethylphenyl)-3-methyl-4-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridine-5-carboxylic acid

  • MDL number MFCD09802209
  • Cas Number : 949328-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87447
Availability details for
Quantity
  • Availability
  • Add to Cart