ENA341292146 Enamine

3-(trifluoromethyl)-5,6,7,8-tetrahydroquinolin-5-one

  • MDL number MFCD09802214
  • Cas Number : 944901-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87451

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87451
Availability details for
Quantity
  • Availability
  • Add to Cart