ENA341292147 Enamine

2-amino-9H-xanthen-9-one

  • MDL number MFCD00100368
  • Cas Number : 20061-68-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87452

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87452
Availability details for
Quantity
  • Availability
  • Add to Cart