ENA341292198 Enamine

(2S)-1-(thiophene-2-carbonyl)pyrrolidine-2-carboxylic acid

  • MDL number MFCD08310544
  • Cas Number : 117918-58-8
  • Molecular Weight : 225.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-87505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87505
Availability details for
Quantity
  • Availability
  • Add to Cart