ENA341292251 Enamine

(2S,4R)-1-(4-fluorobenzenesulfonyl)-4-hydroxypyrrolidine-2-carboxylic acid

  • MDL number MFCD09863512
  • Cas Number : 1613637-25-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87559

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87559
Availability details for
Quantity
  • Availability
  • Add to Cart