ENA341292270 Enamine

(2S)-2-benzenesulfonamido-3-methylbutanoic acid

  • MDL number MFCD09749739
  • Cas Number : 85849-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87578

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87578
Availability details for
Quantity
  • Availability
  • Add to Cart