ENA341292286 Enamine

(2S)-2-benzenesulfonamidopropanoic acid

  • MDL number MFCD08691069
  • Cas Number : 29268-18-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87594

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87594
Availability details for
Quantity
  • Availability
  • Add to Cart