ENA341292321 Enamine

(2S)-2-(naphthalene-2-sulfonamido)-3-phenylpropanoic acid

  • MDL number MFCD02556465
  • Cas Number : 55953-52-1
  • Molecular Weight : 355.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-87629

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87629
Availability details for
Quantity
  • Availability
  • Add to Cart