ENA341292514 Enamine

2-chloro-5-(trifluoromethyl)-1,3-benzothiazole

  • MDL number MFCD09749242
  • Cas Number : 23420-88-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 57...59
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87837

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87837
Availability details for
Quantity
  • Availability
  • Add to Cart