ENA341292518 Enamine

1-(4-hydroxyphenyl)-2-(pyridin-4-yl)ethan-1-one hydrobromide

  • MDL number MFCD20441588
  • Cas Number : 1354954-46-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87841
Availability details for
Quantity
  • Availability
  • Add to Cart