ENA341292546 Enamine

2-(2,6-dichlorophenyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD12068285
  • Cas Number : 1157698-73-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144...146
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87866

Safety Information

msds https://www.enaminestore.com/catalog/EN300-87866
Availability details for
Quantity
  • Availability
  • Add to Cart