ENA341292552 Enamine

2-[2-(6-oxo-1,6-dihydropyridazine-3-amido)-1,3-thiazol-4-yl]acetic acid

  • MDL number MFCD12544311
  • Cas Number : 1098367-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-87872

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87872
Availability details for
Quantity
  • Availability
  • Add to Cart