ENA341292555 Enamine

4-(3-methanesulfonylpropoxy)-2,3-dihydro-1H-inden-1-ol

  • MDL number MFCD18546433
  • Cas Number : 1354951-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87875

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87875
Availability details for
Quantity
  • Availability
  • Add to Cart