ENA341292559 Enamine

2-nitropyridine-3-sulfonyl chloride

  • MDL number MFCD20441589
  • Cas Number : 1354961-20-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87879
Availability details for
Quantity
  • Availability
  • Add to Cart