ENA341292565 Enamine

(1R)-1-[4-(propan-2-yl)phenyl]ethan-1-ol

  • MDL number MFCD09863760
  • Cas Number : 105364-42-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87885

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87885
Availability details for
Quantity
  • Availability
  • Add to Cart