ENA341292590 Enamine

(1S)-1-(5,6,7,8-tetrahydronaphthalen-2-yl)ethan-1-ol

  • MDL number MFCD09863779
  • Cas Number : 1212283-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87911
Availability details for
Quantity
  • Availability
  • Add to Cart