ENA341292636 Enamine

methyl 2-(pyridin-3-yl)propanoate

  • MDL number MFCD16748889
  • Cas Number : 154369-12-7
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-87959

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-87959
Availability details for
Quantity
  • Availability
  • Add to Cart