ENA341292674 Enamine

4-(difluoromethanesulfonylmethyl)benzoic acid

  • MDL number MFCD09863841
  • Cas Number : 1000931-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-88015

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88015
Availability details for
Quantity
  • Availability
  • Add to Cart