ENA341292678 Enamine

2-amino-2-(4-ethylphenyl)acetic acid

  • MDL number MFCD02662425
  • Cas Number : 318270-08-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88019

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88019
Availability details for
Quantity
  • Availability
  • Add to Cart