ENA341292792 Enamine

7,8-dipropoxy-2,3,4,5-tetrahydro-1H-3-benzazepine

  • MDL number MFCD10686952
  • Cas Number : 1050886-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88140
Availability details for
Quantity
  • Availability
  • Add to Cart