ENA341292851 Enamine

N-methyl-1H-imidazole-1-carboxamide

  • MDL number MFCD11099725
  • Cas Number : 72002-25-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 118...120
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88201

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-88201
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart