ENA341292866 Enamine

3-methyl-1-(1,2,3,4-tetrahydroisoquinoline-2-carbonyl)-1H-imidazol-3-ium iodide

  • MDL number MFCD11099740
  • Cas Number : 223762-23-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88216

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88216
Availability details for
Quantity
  • Availability
  • Add to Cart