ENA341292869 Enamine

1-(3,5-dimethylpiperidine-1-carbonyl)-3-methyl-1H-imidazol-3-ium iodide

  • MDL number MFCD11099743
  • Cas Number : 1231954-03-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88219
Availability details for
Quantity
  • Availability
  • Add to Cart