ENA341292877 Enamine

3-methyl-1-[4-(pyridin-2-yl)piperazine-1-carbonyl]-1H-imidazol-3-ium iodide

  • MDL number MFCD11099750
  • Cas Number : 1231959-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data I-
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-88227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-88227
Availability details for
Quantity
  • Availability
  • Add to Cart